Assay Details
Target Gene Details
Entrez Gene ID: | 55870 |
Gene Name: | ASH1 like histone lysine methyltransferase |
Gene Aliases: |
ASH1, ASH1L1, KMT2H |
Location: |
Chr.1:155335261-155563160 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASH1L | NM_018489.2 | 24 | 8698 | NP_060959.2 |
| XM_006711450.3 | 24 | 8389 | XP_006711513.1 | |
| XM_006711451.3 | 25 | 8476 | XP_006711514.1 | |
| XM_006711452.3 | 23 | 8518 | XP_006711515.1 | |
| XM_011509770.2 | 23 | 4069 | XP_011508072.1 | |
| XM_017001784.1 | 24 | 8475 | XP_016857273.1 | |
| XM_017001785.1 | 24 | 8405 | XP_016857274.1 | |
| XM_017001788.1 | 21 | 3193 | XP_016857277.1 | |
| AF257305.1 | 24 | 8542 | AAF68983.1 | |
| AK304873.1 | 5 | 508 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831636 | Chr.1:155249432 - 155475073 on Build GRCh38 | Loss |
|
| esv33869 | Chr.1:155253492 - 155948170 on Build GRCh38 | Loss |
|
| dgv3n68 | Chr.1:155122502 - 155343618 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map