Assay Details
Target Gene Details
Entrez Gene ID: | 1351 |
Gene Name: | cytochrome c oxidase subunit 8A |
Gene Aliases: |
COX, COX8, COX8-2, COX8L, VIII, VIII-L |
Location: |
Chr.11:63974607-63976543 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX8A | NM_004074.2 | 1 | 68 | NP_004065.1 |
| AK311934.1 | ||||
| BC063025.1 | 1 | 68 | AAH63025.1 | |
| BT009803.1 | AAP88805.1 | |||
| CR407687.1 | CAG28615.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion |
|
| nsv1042075 | Chr.11:63964982 - 64045692 on Build GRCh38 | Loss |
|
| esv29993 | Chr.11:63971170 - 64096739 on Build GRCh38 | Loss |
|
| nsv555181 | Chr.11:63872409 - 64015880 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map