Assay Details
Target Gene Details
Entrez Gene ID: | 55139 |
Gene Name: | ankyrin repeat and zinc finger domain containing 1 |
Gene Aliases: |
ZNF744 |
Location: |
Chr.2:219229757-219236669 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKZF1 | NM_001042410.1 | NP_001035869.1 | ||
| NM_001282792.1 | NP_001269721.1 | |||
| NM_018089.2 | NP_060559.2 | |||
| XM_005246663.2 | XP_005246720.1 | |||
| XM_011511392.2 | XP_011509694.1 | |||
| XM_017004420.1 | XP_016859909.1 | |||
| XM_017004421.1 | XP_016859910.1 | |||
| AA482007.1 | ||||
| AF364318.1 | ||||
| AK001277.1 | BAA91596.1 | |||
| AK023206.1 | BAB14462.1 | |||
| AK056060.1 | ||||
| AK095304.1 | ||||
| AK225275.1 | ||||
| AK225309.1 | ||||
| BC000238.1 | AAH00238.1 | |||
| BC008948.2 | AAH08948.1 | |||
| DQ412564.1 | ABD65410.1 |
Target Gene Details
Entrez Gene ID: | 79411 |
Gene Name: | galactosidase beta 1 like |
Gene Aliases: |
- |
Location: |
Chr.2:219236598-219245429 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLB1L | NM_001286423.1 | 17 | 2664 | NP_001273352.1 |
| NM_001286427.1 | 14 | 2181 | NP_001273356.1 | |
| NM_024506.4 | 17 | 2652 | NP_078782.3 | |
| XM_011511813.2 | XP_011510115.1 | |||
| XM_017004895.1 | XP_016860384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834543 | Chr.2:219226398 - 219416430 on Build GRCh38 | Loss |
|
| esv3594357 | Chr.2:219236349 - 219237364 on Build GRCh38 | Loss |
|
| esv3893555 | Chr.2:219172722 - 219273269 on Build GRCh38 | Loss |
|
| nsv508885 | Chr.2:219231225 - 219333602 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map