Assay Details
Target Gene Details
Entrez Gene ID: | 55520 |
Gene Name: | elaC ribonuclease Z 1 |
Gene Aliases: |
D29 |
Location: |
Chr.18:50968017-50988120 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELAC1 | NM_018696.2 | 3 | 672 | NP_061166.1 |
| AB029151.1 | 3 | 612 | BAA96799.1 | |
| AF308695.1 | 3 | 656 | AAG24917.1 | |
| AK024822.1 | 3 | 652 | BAB15021.1 | |
| AK223507.1 | 3 | 616 | BAD97227.1 | |
| AK310911.1 | 3 | 641 | ||
| BC014624.2 | 3 | 633 | AAH14624.1 |
More Information
Set Membership: |
|

Genomic Map