Assay Details
Target Gene Details
Entrez Gene ID: | 63941 |
Gene Name: | N-terminal EF-hand calcium binding protein 3 |
Gene Aliases: |
APBA2BP, EFCBP3, NIP1, STIP3, SYTIP2, XB51, dJ63M2.4, dJ63M2.5 |
Location: |
Chr.20:33657087-33674458 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NECAB3 | NM_031231.3 | 13 | 1345 | NP_112508.3 |
| NM_031232.3 | 12 | 1447 | NP_112509.3 | |
| XM_005260510.1 | 12 | 1261 | XP_005260567.1 | |
| XM_011528991.1 | 9 | 971 | XP_011527293.1 | |
| XM_017028015.1 | 9 | 970 | XP_016883504.1 | |
| XM_017028016.1 | 9 | 899 | XP_016883505.1 | |
| AB039947.1 | 10 | 1149 | BAB16413.1 | |
| AF193759.1 | AAG28415.1 | |||
| AF409141.1 | 13 | 1293 | AAL01118.1 | |
| AK023706.1 | 12 | 1551 | BAB14649.1 | |
| AK124388.1 | 9 | 1419 | ||
| AK291895.1 | 13 | 1275 | ||
| BC047673.1 | 13 | 1300 | AAH47673.1 | |
| CD580591.2 | 2 | 285 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv585927 | Chr.20:33656331 - 33663590 on Build GRCh38 | Loss |
|
| nsv954024 | Chr.20:33638795 - 33698694 on Build GRCh38 | Deletion |
|
| nsv1065189 | Chr.20:33597378 - 33807579 on Build GRCh38 | Loss |
|
| nsv833961 | Chr.20:33605380 - 33836992 on Build GRCh38 | Loss |
|
| nsv833960 | Chr.20:33569059 - 33688060 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map