Assay Details
Target Gene Details
Entrez Gene ID: | 10318 |
Gene Name: | TNFAIP3 interacting protein 1 |
Gene Aliases: |
ABIN-1, NAF1, VAN, nip40-1 |
Location: |
Chr.5:151029943-151087660 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNIP1 | NM_001252385.1 | 18 | 2736 | NP_001239314.1 |
| NM_001252386.1 | 17 | 2547 | NP_001239315.1 | |
| NM_001252390.1 | 18 | 2584 | NP_001239319.1 | |
| NM_001252391.1 | 18 | 2616 | NP_001239320.1 | |
| NM_001252392.1 | 18 | 2542 | NP_001239321.1 | |
| NM_001252393.1 | 18 | 2645 | NP_001239322.1 | |
| NM_001258454.1 | 18 | 2719 | NP_001245383.1 | |
| NM_001258455.1 | 16 | 2322 | NP_001245384.1 | |
| NM_001258456.1 | 15 | 2225 | NP_001245385.1 | |
| NM_006058.4 | 18 | 2719 | NP_006049.3 | |
| XM_005268355.1 | 18 | 2575 | XP_005268412.1 | |
| XM_006714751.1 | 17 | 2440 | XP_006714814.1 | |
| XM_017008951.1 | 17 | 2819 | XP_016864440.1 | |
| AJ011895.1 | 18 | 2588 | CAA09855.1 | |
| AJ011896.1 | 18 | 2514 | CAA09856.1 | |
| AK074400.1 | 18 | 2650 | ||
| AK097213.1 | 1 | 2711 | ||
| AK303573.1 | 18 | 2686 | ||
| AW589284.1 | 1 | 215 | ||
| AY012155.1 | 18 | 2735 | AAG42154.1 | |
| BC012133.1 | 18 | 2625 | AAH12133.1 | |
| BC014008.1 | 18 | 2566 | AAH14008.1 | |
| BX640647.1 | 17 | 2468 | CAE45793.1 | |
| D30755.2 | 17 | 2478 | BAA06416.2 | |
| U39403.1 | 14 | 2000 | AAC99999.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526224 | Chr.5:151003841 - 151070675 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map