Assay Details
Target Gene Details
Entrez Gene ID: | 340527 |
Gene Name: | NHS like 2 |
Gene Aliases: |
- |
Location: |
Chr.X:71911073-72153286 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NHSL2 | NM_001013627.2 | NP_001013649.2 | ||
| XM_011530932.1 | XP_011529234.1 | |||
| XM_011530933.1 | XP_011529235.1 | |||
| XM_011530934.2 | XP_011529236.1 | |||
| XM_017029473.1 | XP_016884962.1 | |||
| XM_017029474.1 | XP_016884963.1 | |||
| XM_017029475.1 | XP_016884964.1 | |||
| XM_017029476.1 | XP_016884965.1 | |||
| XM_017029477.1 | XP_016884966.1 |
Target Gene Details
Entrez Gene ID: | 340526 |
Gene Name: | retrotransposon gag domain containing 4 |
Gene Aliases: |
6430402L03Rik, MAR5, MART5 |
Location: |
Chr.X:72127110-72131901 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGAG4 | NM_001024455.3 | 2 | 4259 | NP_001019626.1 |
| AB082532.1 | 2 | 4259 | ||
| AL359579.2 | 2 | 3303 | CAB94872.2 | |
| BC151224.1 | 2 | 4280 |
More Information
Set Membership: |
|

Genomic Map