Assay Details
Target Gene Details
Entrez Gene ID: | 10436 |
Gene Name: | EMG1, N1-specific pseudouridine methyltransferase |
Gene Aliases: |
C2F, Grcc2f, NEP1 |
Location: |
Chr.12:6970781-6997428 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EMG1 | NM_001320049.1 | 5 | 783 | NP_001306978.1 |
| NM_006331.7 | 6 | 933 | NP_006322.4 | |
| NR_135131.1 | ||||
| AI394112.1 | 1 | 140 | ||
| AK291597.1 | 6 | 798 | ||
| AK298745.1 | ||||
| AK307177.1 | 3 | 566 | ||
| BC055314.1 | 6 | 825 | AAH55314.1 | |
| BQ015307.1 | 1 | 136 | ||
| U72514.1 | 6 | 776 | AAC51641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047373 | Chr.12:6902891 - 7055754 on Build GRCh38 | Gain |
|
| nsv509453 | Chr.12:6896530 - 7007852 on Build GRCh38 | Insertion |
|
| nsv1035811 | Chr.12:6876316 - 6994911 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map