Assay Details
Target Gene Details
Entrez Gene ID: | 10000 |
Gene Name: | AKT serine/threonine kinase 3 |
Gene Aliases: |
MPPH, MPPH2, PKB-GAMMA, PKBG, PRKBG, RAC-PK-gamma, RAC-gamma, STK-2 |
Location: |
Chr.1:243488233-243851079 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKT3 | NM_001206729.1 | NP_001193658.1 | ||
| NM_005465.4 | 13 | 2064 | NP_005456.1 | |
| NM_181690.2 | NP_859029.1 | |||
| XM_005272994.4 | 14 | 2289 | XP_005273051.1 | |
| XM_005272995.2 | 14 | 2095 | XP_005273052.1 | |
| XM_011544014.2 | 7 | 1472 | XP_011542316.1 | |
| XM_016999985.1 | 15 | 2237 | XP_016855474.1 | |
| AF135794.1 | 13 | 1952 | AAD24196.1 | |
| AL117525.1 | CAB55977.1 | |||
| AY005799.1 | AAF91073.1 | |||
| BC121154.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833070 | Chr.1:243339809 - 243537501 on Build GRCh38 | Gain |
|
| nsv1010479 | Chr.1:243494728 - 243515087 on Build GRCh38 | Loss |
|
| dgv601n100 | Chr.1:243494646 - 243515087 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map