Assay Details
Target Gene Details
Entrez Gene ID: | 7170 |
Gene Name: | tropomyosin 3 |
Gene Aliases: |
CAPM1, CFTD, HEL-189, HEL-S-82p, NEM1, OK/SW-cl.5, TM-5, TM3, TM30, TM30nm, TM5, TPMsk3, TRK, hscp30 |
Location: |
Chr.1:154155304-154192135 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPM3 | NM_001043351.1 | 8 | 3082 | NP_001036816.1 |
| NM_001043352.1 | 8 | 4409 | NP_001036817.1 | |
| NM_001043353.1 | 8 | 4409 | NP_001036818.1 | |
| NM_001278188.1 | 7 | 2948 | NP_001265117.1 | |
| NM_001278189.1 | 9 | 3161 | NP_001265118.1 | |
| NM_001278190.1 | 7 | 3019 | NP_001265119.1 | |
| NM_001278191.1 | 8 | 2928 | NP_001265120.1 | |
| NM_153649.3 | 8 | 3082 | NP_705935.1 | |
| NR_103461.1 | 8 | 3078 | ||
| AA863064.1 | 1 | 134 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520398 | Chr.1:154095720 - 154169432 on Build GRCh38 | Gain+Loss |
|
| nsv1000153 | Chr.1:154101773 - 154179941 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map