Assay Details
Target Gene Details
Entrez Gene ID: | 51360 |
Gene Name: | membrane bound transcription factor peptidase, site 2 |
Gene Aliases: |
BRESEK, IFAP, KFSD, KFSDX, OLMSX, S2P |
Location: |
Chr.X:21839538-21885423 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBTPS2 | NM_015884.3 | NP_056968.1 | ||
| AF019612.1 | AAC51937.1 | |||
| AK292933.1 | ||||
| AK307143.1 | ||||
| AK312474.1 |
Target Gene Details
Entrez Gene ID: | 404281 |
Gene Name: | YY2 transcription factor |
Gene Aliases: |
ZNF631 |
Location: |
Chr.X:21855987-21858727 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YY2 | NM_206923.3 | 1 | 544 | NP_996806.2 |
| AK091850.1 | 1 | 544 | ||
| AY567472.1 | 1 | 46 | AAS68634.1 | |
| BC137215.1 | 1 | 100 | ||
| BC137217.1 | 1 | 100 | ||
| DA961884.1 | 1 | 70 | ||
| DB029513.1 | 1 | 296 | ||
| DB459498.2 |
More Information
Set Membership: |
|

Genomic Map