Assay Details
Target Gene Details
Entrez Gene ID: | 56937 |
Gene Name: | prostate transmembrane protein, androgen induced 1 |
Gene Aliases: |
STAG1, TMEPAI |
Location: |
Chr.20:57648392-57711536 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PMEPA1 | NM_001255976.1 | 4 | 4296 | NP_001242905.1 |
| NM_020182.4 | 4 | 4567 | NP_064567.2 | |
| NM_199169.2 | 4 | 4220 | NP_954638.1 | |
| NM_199170.2 | 4 | 4213 | NP_954639.1 | |
| NM_199171.2 | 4 | 4278 | NP_954640.1 | |
| AF305616.1 | 4 | 4494 | AAL16781.1 | |
| AK024618.1 | 1 | 1442 | ||
| BQ008707.1 | 1 | 368 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758802 | Chr.20:57595722 - 58113215 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112660826,rs201737117] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map