Assay Details
Target Gene Details
Entrez Gene ID: | 51490 |
Gene Name: | SPOUT domain containing methyltransferase 1 |
Gene Aliases: |
C9orf114, CENP-32, HSPC109 |
Location: |
Chr.9:128819651-128833309 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPOUT1 | NM_016390.3 | NP_057474.2 | ||
| XM_017014804.1 | XP_016870293.1 | |||
| AF161459.1 | AAF29074.1 | |||
| AK303563.1 | ||||
| AL110193.1 | ||||
| BC010579.1 | AAH10579.1 | |||
| BC021273.1 | ||||
| BC033677.1 | AAH33677.1 | |||
| BC039590.1 | AAH39590.1 | |||
| BC046133.1 | AAH46133.1 | |||
| BC063644.1 | AAH63644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6724 | Chr.9:128823206 - 128858415 on Build GRCh38 | Insertion |
|
| nsv521120 | Chr.9:128803275 - 128829871 on Build GRCh38 | Loss |
|
| nsv615411 | Chr.9:128727172 - 128829871 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map