Assay Details
Target Gene Details
Entrez Gene ID: | 8892 |
Gene Name: | eukaryotic translation initiation factor 2B subunit beta |
Gene Aliases: |
EIF-2Bbeta, EIF2B |
Location: |
Chr.14:75002909-75009591 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2B2 | NM_014239.3 | NP_055054.1 | ||
| AF035280.1 | AAB88176.1 | |||
| BC000494.2 | AAH00494.1 | |||
| BC003165.1 | AAH03165.1 | |||
| BC011750.2 | AAH11750.1 | |||
| BI908906.1 | ||||
| BX161473.1 | CAD61929.1 | |||
| L40395.1 | AAC42002.1 |
More Information
Set Membership: |
|

Genomic Map