Assay Details
Target Gene Details
Entrez Gene ID: | 22854 |
Gene Name: | netrin G1 |
Gene Aliases: |
Lmnt1 |
Location: |
Chr.1:107139918-107484899 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTNG1 | NM_001113226.2 | 1 | 327 | NP_001106697.1 |
| NM_001113228.2 | 1 | 327 | NP_001106699.1 | |
| NM_001312688.1 | 2 | 609 | NP_001299617.1 | |
| NM_014917.3 | 2 | 609 | NP_055732.2 | |
| XM_006710455.2 | 2 | 1243 | XP_006710518.1 | |
| XM_006710456.3 | 2 | 1243 | XP_006710519.1 | |
| XM_011541021.1 | 2 | 1243 | XP_011539323.1 | |
| XM_011541024.2 | 2 | 1243 | XP_011539326.1 | |
| XM_011541025.1 | 2 | 1243 | XP_011539327.1 | |
| XM_017000680.1 | 2 | 609 | XP_016856169.1 | |
| XM_017000681.1 | 2 | 1562 | XP_016856170.1 | |
| XM_017000682.1 | 2 | 1243 | XP_016856171.1 | |
| XM_017000683.1 | 2 | 609 | XP_016856172.1 | |
| XM_017000684.1 | 2 | 1562 | XP_016856173.1 | |
| XM_017000685.1 | 2 | 1562 | XP_016856174.1 | |
| XM_017000686.1 | 2 | 510 | XP_016856175.1 | |
| AB023193.1 | 2 | 541 | BAA76820.2 | |
| AK296533.1 | 2 | 510 | ||
| AY358365.1 | 1 | 67 | AAQ88731.1 | |
| BC030220.1 | 2 | 404 | AAH30220.1 | |
| BX538348.1 | 2 | 520 | CAD98143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758957 | Chr.1:106941006 - 107298261 on Build GRCh38 | Loss |
|
| nsv1001678 | Chr.1:107117748 - 107155038 on Build GRCh38 | Loss |
|
| nsv830982 | Chr.1:107108954 - 107292352 on Build GRCh38 | Gain |
|
| nsv547330 | Chr.1:107006123 - 107375697 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map