Assay Details
Target Gene Details
Entrez Gene ID: | 2028 |
Gene Name: | glutamyl aminopeptidase |
Gene Aliases: |
APA, CD249, gp160 |
Location: |
Chr.4:110476073-110563337 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENPEP | NM_001977.3 | 1 | 932 | NP_001968.3 |
| AK313412.1 | 1 | 594 | ||
| AM888273.1 | 1 | 932 | ||
| AM888274.1 | 1 | 932 | ||
| AM888275.1 | 1 | 932 | ||
| AM888276.1 | 1 | 932 | ||
| AM888277.1 | 1 | 932 | ||
| AM888278.1 | 1 | 932 | ||
| AM888279.1 | 1 | 932 | ||
| AM888280.1 | 1 | 932 | ||
| AM888281.1 | 1 | 932 | ||
| AM888282.1 | 1 | 932 | ||
| AM888283.1 | 1 | 932 | ||
| BC094770.1 | 1 | 913 | AAH94770.1 | |
| BX641012.1 | 2 | 702 | ||
| L12468.1 | 1 | 836 | AAA16876.1 | |
| L14721.1 | 1 | 674 | AAA35522.1 |
More Information
Set Membership: |
|

Genomic Map