Assay Details
Target Gene Details
Entrez Gene ID: | 90273 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 21 |
Gene Aliases: |
CEACAM3, R29124_1 |
Location: |
Chr.19:41549518-41586844 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM21 | NM_001098506.3 | NP_001091976.3 | ||
| NM_001288773.2 | NP_001275702.2 | |||
| NM_001290113.1 | NP_001277042.1 | |||
| NM_033543.5 | NP_291021.4 | |||
| XM_005278396.4 | XP_005278453.1 | |||
| XM_005278397.3 | XP_005278454.1 | |||
| XM_011527450.1 | XP_011525752.1 | |||
| XM_011527452.1 | XP_011525754.1 | |||
| XM_017027430.1 | XP_016882919.1 | |||
| XM_017027431.1 | XP_016882920.1 | |||
| XM_017027432.1 | XP_016882921.1 | |||
| XM_017027433.1 | XP_016882922.1 | |||
| XM_017027434.1 | XP_016882923.1 | |||
| XM_017027435.1 | XP_016882924.1 | |||
| XM_017027436.1 | XP_016882925.1 | |||
| XM_017027437.1 | XP_016882926.1 | |||
| AK023602.1 | ||||
| AY358084.1 | AAQ88451.1 | |||
| BC012001.1 | AAH12001.1 | |||
| BC106727.1 | AAI06728.1 | |||
| BI833768.1 | ||||
| BI913326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2718577 | Chr.19:41576456 - 41709018 on Build GRCh38 | Deletion |
|
| nsv519408 | Chr.19:41559909 - 41667978 on Build GRCh38 | Gain |
|
| dgv3549n100 | Chr.19:41550966 - 41671232 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map