Assay Details
Target Gene Details
Entrez Gene ID: | 718 |
Gene Name: | complement component 3 |
Gene Aliases: |
AHUS5, ARMD9, ASP, C3a, C3b, CPAMD1, HEL-S-62p |
Location: |
Chr.19:6677835-6720682 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 39 - Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3 | NM_000064.3 | NP_000055.2 | ||
| AI758500.1 | ||||
| AK094728.1 | ||||
| AK304071.1 | ||||
| BC150179.1 | ||||
| BC150200.1 | ||||
| EU794602.1 | ||||
| K02765.1 | AAA85332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6235n54 | Chr.19:6677148 - 6679405 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map