Assay Details
Target Gene Details
Entrez Gene ID: | 3067 |
Gene Name: | histidine decarboxylase |
Gene Aliases: |
- |
Location: |
Chr.15:50241945-50266049 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDC | NM_001306146.1 | 11 | 2021 | NP_001293075.1 |
| NM_002112.3 | 12 | 2120 | NP_002103.2 | |
| XM_017022094.1 | 12 | 1989 | XP_016877583.1 | |
| XM_017022095.1 | 11 | 1890 | XP_016877584.1 | |
| XM_017022096.1 | 11 | 1774 | XP_016877585.1 | |
| XM_017022097.1 | 10 | 2072 | XP_016877586.1 | |
| XM_017022098.1 | 9 | 1592 | XP_016877587.1 | |
| AK027221.1 | 5 | 1523 | ||
| BC130527.1 | 12 | 1821 | ||
| BC144173.1 | 11 | 1722 | ||
| M60445.1 | 12 | 1829 | AAC41698.1 | |
| X54297.1 | 12 | 1880 | CAA38196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1531 | Chr.15:50225808 - 50257078 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map