Assay Details
Target Gene Details
Entrez Gene ID: | 5700 |
Gene Name: | proteasome 26S subunit, ATPase 1 |
Gene Aliases: |
P26S4, S4, p56 |
Location: |
Chr.14:90256550-90272625 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMC1 | NM_002802.2 | NP_002793.2 | ||
| XM_017021470.1 | XP_016876959.1 | |||
| XM_017021471.1 | XP_016876960.1 | |||
| AF401213.1 | ||||
| AK124201.1 | ||||
| L02426.1 | AAA35484.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv974363 | Chr.14:90271786 - 90272659 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map