Assay Details
Target Gene Details
Entrez Gene ID: | 26038 |
Gene Name: | chromodomain helicase DNA binding protein 5 |
Gene Aliases: |
CHD-5 |
Location: |
Chr.1:6101787-6180134 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 35 - Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHD5 | NM_015557.2 | NP_056372.1 | ||
| AB007913.1 | BAA32289.1 | |||
| AF425231.1 | AAL98962.1 | |||
| AK091386.1 | ||||
| AK126012.1 | BAC86391.1 | |||
| AK127046.1 | BAC86802.1 | |||
| AL110281.1 | CAH56404.1 | |||
| AL117491.1 | CAB55959.1 | |||
| AL137693.1 | CAH56377.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470689 | Chr.1:6089062 - 6173513 on Build GRCh38 | Loss |
|
| nsv461283 | Chr.1:6048317 - 6124032 on Build GRCh38 | Loss |
|
| dgv75n54 | Chr.1:6081265 - 6144206 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map