Assay Details
Target Gene Details
Entrez Gene ID: | 10544 |
Gene Name: | protein C receptor |
Gene Aliases: |
CCCA, CCD41, EPCR |
Location: |
Chr.20:35171937-35215989 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROCR | NM_006404.4 | 1 | 88 | NP_006395.2 |
| XM_011528496.1 | XP_011526798.1 | |||
| AK314887.1 | 1 | 54 | ||
| BC014451.1 | AAH14451.1 | |||
| BG423815.1 | 1 | 55 | ||
| BM808358.1 | 1 | 106 | ||
| HY110044.1 | 1 | 87 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645712 | Chr.20:35167207 - 35174665 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map