Assay Details
Target Gene Details
Entrez Gene ID: | 5650 |
Gene Name: | kallikrein related peptidase 7 |
Gene Aliases: |
PRSS6, SCCE, hK7 |
Location: |
Chr.19:50976479-50984064 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK7 | NM_001207053.1 | 5 | 1901 | NP_001193982.1 |
| NM_001243126.1 | 5 | 1884 | NP_001230055.1 | |
| NM_005046.3 | 6 | 2032 | NP_005037.1 | |
| NM_139277.2 | 6 | 1862 | NP_644806.1 | |
| AI049783.1 | 1 | 73 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064207 | Chr.19:50971269 - 50994022 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map