Assay Details
Target Gene Details
Entrez Gene ID: | 216 |
Gene Name: | aldehyde dehydrogenase 1 family member A1 |
Gene Aliases: |
ALDC, ALDH-E1, ALDH1, ALDH11, HEL-9, HEL-S-53e, HEL12, PUMB1, RALDH1 |
Location: |
Chr.9:72900662-72953317 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH1A1 | NM_000689.4 | 13 | 1840 | NP_000680.2 |
| AB209821.1 | 13 | 1567 | BAD93058.1 | |
| AF003341.1 | AAC51652.1 | |||
| AI074764.1 | ||||
| AK000118.1 | 13 | 1567 | ||
| AK026641.1 | 13 | 1567 | ||
| AK293176.1 | 13 | 1533 | ||
| AK302168.1 | 12 | 1295 | ||
| AY390731.1 | AAR92229.1 | |||
| BC001505.2 | 13 | 1548 | AAH01505.1 | |
| BT006921.1 | AAP35567.1 | |||
| EU668318.1 | 13 | 1567 | ||
| EU794589.1 | ||||
| FJ224286.1 | 13 | 1576 | ||
| K03000.1 | 9 | 1040 | AAA51695.1 | |
| M26761.1 | AAA35518.1 |
More Information
Set Membership: |
|

Genomic Map