Assay Details
Target Gene Details
Entrez Gene ID: | 5705 |
Gene Name: | proteasome 26S subunit, ATPase 5 |
Gene Aliases: |
S8, SUG-1, SUG1, TBP10, TRIP1, p45, p45/SUG |
Location: |
Chr.17:63827410-63832027 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMC5 | NM_001199163.1 | NP_001186092.1 | ||
| NM_002805.5 | NP_002796.4 | |||
| AK095836.1 | ||||
| AK290758.1 | ||||
| AK291878.1 | ||||
| BC001932.1 | AAH01932.1 | |||
| BC002367.2 | AAH02367.3 | |||
| D44467.1 | BAA07919.1 | |||
| L38810.1 | AAC41735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833510 | Chr.17:63767293 - 63927790 on Build GRCh38 | Loss |
|
| nsv517272 | Chr.17:63814307 - 64020132 on Build GRCh38 | Gain+Loss |
|
| nsv510720 | Chr.17:63809351 - 63908780 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map