Assay Details
Target Gene Details
Entrez Gene ID: | 23022 |
Gene Name: | palladin, cytoskeletal associated protein |
Gene Aliases: |
CGI-151, CGI151, MYN, PNCA1, SIH002 |
Location: |
Chr.4:168497039-168928457 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PALLD | NM_001166108.1 | 2 | 950 | NP_001159580.1 |
| NM_016081.3 | 2 | 950 | NP_057165.3 | |
| XM_005262861.4 | 2 | 977 | XP_005262918.1 | |
| XM_011531768.2 | 3 | 1231 | XP_011530070.1 | |
| XM_011531769.2 | 3 | 1231 | XP_011530071.1 | |
| XM_011531770.2 | 3 | 1231 | XP_011530072.1 | |
| XM_011531771.2 | 3 | 1231 | XP_011530073.1 | |
| XM_011531772.2 | 3 | 1231 | XP_011530074.1 | |
| XM_011531773.1 | 3 | 1231 | XP_011530075.1 | |
| XM_011531774.1 | 3 | 1231 | XP_011530076.1 | |
| XM_017007910.1 | 3 | 1231 | XP_016863399.1 | |
| XM_017007911.1 | 3 | 1231 | XP_016863400.1 | |
| AF464873.1 | 2 | 950 | AAL69964.1 | |
| AK096458.1 | 1 | 676 | BAC04796.1 | |
| BC140850.1 | 2 | 912 | ||
| BC144666.1 | 2 | 912 |
More Information
Set Membership: |
|

Genomic Map