Assay Details
Target Gene Details
Entrez Gene ID: | 2193 |
Gene Name: | phenylalanyl-tRNA synthetase alpha subunit |
Gene Aliases: |
CML33, FARSL, FARSLA, FRSA, PheHA |
Location: |
Chr.19:12922470-12933744 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FARSA | NM_004461.2 | NP_004452.1 | ||
| AF042347.1 | AAD02221.1 | |||
| AK092093.1 | ||||
| AK130195.1 | ||||
| AK304587.1 | ||||
| BC006495.2 | AAH06495.1 | |||
| BC043565.1 | AAH43565.1 | |||
| BT007198.1 | AAP35862.1 | |||
| CR456740.1 | CAG33021.1 | |||
| D84471.1 | BAA95666.1 | |||
| U07424.1 | AAB61694.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160589 | Chr.19:12898242 - 12948155 on Build GRCh38 | Duplication |
|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|
| nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map