Assay Details
Target Gene Details
Entrez Gene ID: | 8527 |
Gene Name: | diacylglycerol kinase delta |
Gene Aliases: |
DGKdelta, dgkd-2 |
Location: |
Chr.2:233354507-233472098 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DGKD | NM_003648.2 | 1 | 53 | NP_003639.2 |
| NM_152879.2 | NP_690618.2 | |||
| XM_011512028.2 | XP_011510330.1 | |||
| XM_011512029.2 | XP_011510331.1 | |||
| XM_011512030.2 | XP_011510332.1 | |||
| XM_011512031.2 | XP_011510333.1 | |||
| XM_011512032.2 | XP_011510334.1 | |||
| XM_011512033.2 | XP_011510335.1 | |||
| XM_011512039.2 | XP_011510341.1 | |||
| XM_017005138.1 | XP_016860627.1 | |||
| XM_017005140.1 | XP_016860629.1 | |||
| AB078966.1 | BAC11809.1 | |||
| D63479.3 | BAA09766.3 | |||
| D73409.1 | 1 | 54 | BAA11134.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584700 | Chr.2:233290755 - 233455780 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map