Assay Details
Target Gene Details
Entrez Gene ID: | 7040 |
Gene Name: | transforming growth factor beta 1 |
Gene Aliases: |
CED, DPD1, LAP, TGFB, TGFbeta |
Location: |
Chr.19:41330531-41353933 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFB1 | NM_000660.5 | 3 | 1471 | NP_000651.3 |
| XM_011527242.1 | 3 | 1464 | XP_011525544.1 | |
| AK291907.1 | 3 | 960 | ||
| AK307742.1 | 3 | 1156 | ||
| AY820829.1 | 2 | 85 | AAV71148.1 | |
| BC000125.1 | 3 | 1028 | AAH00125.1 | |
| BC001180.1 | 3 | 1028 | AAH01180.1 | |
| BC022242.1 | 3 | 951 | AAH22242.1 | |
| BT007245.1 | 3 | 582 | AAP35909.1 | |
| X02812.1 | 3 | 1426 | CAA26580.1 |
More Information
Set Membership: |
|

Genomic Map