Assay Details
Target Gene Details
Entrez Gene ID: | 7277 |
Gene Name: | tubulin alpha 4a |
Gene Aliases: |
ALS22, H2-ALPHA, TUBA1 |
Location: |
Chr.2:219249711-219254608 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBA4A | NM_001278552.1 | NP_001265481.1 | ||
| NM_006000.2 | NP_005991.1 | |||
| XM_017004824.1 | XP_016860313.1 | |||
| AK054731.1 | ||||
| AK299958.1 | ||||
| BC009238.2 | AAH09238.1 | |||
| BP346611.1 |
Target Gene Details
Entrez Gene ID: | 80086 |
Gene Name: | tubulin alpha 4b |
Gene Aliases: |
TUBA4 |
Location: |
Chr.2:219253243-219272188 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBA4B | NR_003063.1 | |||
| AK024002.1 | BAB14767.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834543 | Chr.2:219226398 - 219416430 on Build GRCh38 | Loss |
|
| esv3893555 | Chr.2:219172722 - 219273269 on Build GRCh38 | Loss |
|
| nsv954842 | Chr.2:219250279 - 219254378 on Build GRCh38 | Deletion |
|
| nsv508885 | Chr.2:219231225 - 219333602 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map