Assay Details
Target Gene Details
Entrez Gene ID: | 148327 |
Gene Name: | cAMP responsive element binding protein 3 like 4 |
Gene Aliases: |
AIBZIP, ATCE1, CREB3, CREB4, JAL, hJAL |
Location: |
Chr.1:153967679-153974364 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB3L4 | NM_001255978.1 | 10 | 1677 | NP_001242907.1 |
| NM_001255979.1 | 10 | 1463 | NP_001242908.1 | |
| NM_001255980.1 | 10 | 1617 | NP_001242909.1 | |
| NM_001255981.1 | 10 | 1403 | NP_001242910.1 | |
| NM_130898.3 | 10 | 1697 | NP_570968.1 | |
| NR_045658.1 | 9 | 1430 | ||
| XM_006711172.2 | 10 | 1585 | XP_006711235.1 | |
| XM_017000372.1 | 10 | 1375 | XP_016855861.1 | |
| AA652478.1 | 1 | 153 | ||
| AB052778.1 | 10 | 1615 | BAC45035.1 | |
| AF394167.1 | 10 | 1425 | AAL76113.1 | |
| AF468007.1 | 10 | 1464 | AAO33070.1 | |
| AK297703.1 | 9 | 1399 | ||
| AK304665.1 | 10 | 1542 | ||
| AY049977.1 | 10 | 1613 | AAL13157.1 | |
| BC038962.1 | 10 | 1599 | AAH38962.1 | |
| BX647716.1 | 10 | 1362 |
Target Gene Details
Entrez Gene ID: | 10899 |
Gene Name: | jumping translocation breakpoint |
Gene Aliases: |
HJTB, HSPC222, PAR, hJT |
Location: |
Chr.1:153974269-153977975 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JTB | NM_006694.3 | NP_006685.1 | ||
| AF131797.1 | AAD20045.1 | |||
| BC000996.2 | AAH00996.1 |
More Information
Set Membership: |
|

Genomic Map