Assay Details
Target Gene Details
Entrez Gene ID: | 6697 |
Gene Name: | sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase) |
Gene Aliases: |
SDR38C1 |
Location: |
Chr.2:72887383-72892160 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPR | NM_003124.4 | NP_003115.1 | ||
| AK222942.1 | BAD96662.1 | |||
| AK291856.1 | ||||
| BC017310.2 | AAH17310.1 | |||
| M76231.1 | AAA60314.1 | |||
| S71376.1 | AAD14079.1 |
More Information
Set Membership: |
|

Genomic Map