Assay Details
Target Gene Details
Entrez Gene ID: | 7716 |
Gene Name: | vascular endothelial zinc finger 1 |
Gene Aliases: |
DB1, ZNF161 |
Location: |
Chr.17:57971547-57988254 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VEZF1 | NM_007146.2 | 6 | 4556 | NP_009077.2 |
| XM_005257643.1 | 6 | 4553 | XP_005257700.1 | |
| XM_005257644.4 | 7 | 4709 | XP_005257701.1 | |
| XM_017025017.1 | 7 | 4760 | XP_016880506.1 | |
| XM_017025018.1 | 7 | 4612 | XP_016880507.1 | |
| AK058024.1 | 1 | 886 | ||
| BX648862.1 | 6 | 3849 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833497 | Chr.17:57929210 - 58108015 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map