Assay Details
Target Gene Details
Entrez Gene ID: | 5267 |
Gene Name: | serpin family A member 4 |
Gene Aliases: |
KAL, KLST, KST, PI-4, PI4, kallistatin |
Location: |
Chr.14:94561420-94569913 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINA4 | NM_001289032.1 | 2 | 506 | NP_001275961.1 |
| NM_001289033.1 | 2 | 351 | NP_001275962.1 | |
| NM_006215.3 | 2 | 191 | NP_006206.2 | |
| AK309586.1 | 2 | 484 | ||
| AK313196.1 | 2 | 169 | ||
| BC014992.1 | 2 | 165 | AAH14992.1 | |
| BX248009.1 | 2 | 266 | CAD62337.1 | |
| BX248760.1 | 2 | 154 | CAD66567.1 | |
| DA036536.1 | 2 | 191 | ||
| DA637776.1 | 2 | 296 | ||
| L19684.1 | 1 | 99 | AAA59454.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470662 | Chr.14:94296542 - 94798793 on Build GRCh38 | Gain |
|
| nsv565623 | Chr.14:94393116 - 94635006 on Build GRCh38 | Gain |
|
| nsv832865 | Chr.14:94499457 - 94667894 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map