Assay Details
Target Gene Details
Entrez Gene ID: | 124637 |
Gene Name: | cytochrome b5 domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.17:7857746-7864417 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB5D1 | NM_144607.4 | 1 | 67 | NP_653208.2 |
| AK057061.1 | 1 | 67 | BAB71357.1 |
Target Gene Details
Entrez Gene ID: | 84316 |
Gene Name: | N(alpha)-acetyltransferase 38, NatC auxiliary subunit |
Gene Aliases: |
LSMD1, PFAAP2 |
Location: |
Chr.17:7856685-7885465 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA38 | NM_001320924.1 | 1 | 80 | NP_001307853.1 |
| NM_001320925.1 | 1 | 80 | NP_001307854.1 | |
| NM_032356.4 | 1 | 80 | NP_115732.2 | |
| XM_011524037.2 | XP_011522339.1 | |||
| XM_017025225.1 | XP_016880714.1 | |||
| XM_017025226.1 | XP_016880715.1 | |||
| BC033861.1 | 1 | 43 | AAH33861.1 | |
| BC051846.1 | 1 | 80 | AAH51846.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv510n67 | Chr.17:7852429 - 7911448 on Build GRCh38 | Gain |
|
| nsv833351 | Chr.17:7755691 - 7893863 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs150998226] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map