Assay Details
Target Gene Details
Entrez Gene ID: | 5337 |
Gene Name: | phospholipase D1 |
Gene Aliases: |
- |
Location: |
Chr.3:171600404-171810494 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLD1 | NM_001130081.2 | 26 | 5224 | NP_001123553.1 |
| NM_002662.4 | 27 | 5338 | NP_002653.1 | |
| XM_005247533.2 | 27 | 5407 | XP_005247590.1 | |
| XM_005247534.2 | 26 | 5293 | XP_005247591.1 | |
| XM_011512899.2 | 17 | 4490 | XP_011511201.1 | |
| XM_017006624.1 | 16 | 4612 | XP_016862113.1 | |
| AB209907.1 | 26 | 5168 | BAD93144.1 | |
| BC068976.1 | 27 | 5289 | AAH68976.1 | |
| BX648900.1 | 13 | 3753 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001697 | Chr.3:171589803 - 171775069 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map