Assay Details
Target Gene Details
Entrez Gene ID: | 57709 |
Gene Name: | solute carrier family 7 member 14 |
Gene Aliases: |
PPP1R142 |
Location: |
Chr.3:170459554-170586074 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A14 | NM_020949.2 | 8 | 2877 | NP_066000.2 |
| AB046833.1 | 7 | 2328 | BAB13439.1 | |
| AK122655.1 | ||||
| BC022968.1 |
More Information
Set Membership: |
|

Genomic Map