Assay Details
Target Gene Details
Entrez Gene ID: | 4142 |
Gene Name: | MAS1 proto-oncogene, G protein-coupled receptor |
Gene Aliases: |
MAS, MGRA |
Location: |
Chr.6:159906942-159908076 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAS1 | NM_002377.2 | 1 | 932 | NP_002368.1 |
| BC069142.1 | 1 | 933 | AAH69142.1 | |
| BC069581.1 | 1 | 932 | AAH69581.1 | |
| BC110454.2 | 1 | 932 | AAI10455.1 | |
| CR542261.1 | CAG47057.1 | |||
| M13150.1 | 1 | 1184 | AAA36199.1 |
More Information
Set Membership: |
|

Genomic Map