Assay Details
Target Gene Details
Entrez Gene ID: | 55168 |
Gene Name: | mitochondrial ribosomal protein S18A |
Gene Aliases: |
HumanS18b, MRP-S18-3, MRPS18-3, S18bmt |
Location: |
Chr.6:43671197-43687812 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS18A | NM_001193343.1 | NP_001180272.1 | ||
| NM_018135.3 | NP_060605.1 | |||
| XM_006715134.2 | XP_006715197.1 | |||
| AB049952.1 | BAB41005.1 | |||
| AK001410.1 | BAA91675.1 | |||
| AK300757.1 | ||||
| AK307183.1 | ||||
| BC010364.1 | AAH10364.1 | |||
| DQ884400.1 |
More Information
Set Membership: |
|

Genomic Map