Assay Details
Target Gene Details
Entrez Gene ID: | 80975 |
Gene Name: | transmembrane protease, serine 5 |
Gene Aliases: |
SPINESIN |
Location: |
Chr.11:113687546-113709811 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS5 | NM_001288749.1 | 5 | 571 | NP_001275678.1 |
| NM_001288750.1 | 5 | 571 | NP_001275679.1 | |
| NM_001288751.1 | 6 | 778 | NP_001275680.1 | |
| NM_001288752.1 | 6 | 674 | NP_001275681.1 | |
| NM_030770.3 | 6 | 674 | NP_110397.2 | |
| NR_110046.1 | 4 | 472 | ||
| NR_110047.1 | 4 | 472 | ||
| XM_017018366.1 | 4 | 3910 | XP_016873855.1 | |
| XM_017018367.1 | 6 | 4102 | XP_016873856.1 | |
| AB028140.1 | 6 | 680 | BAB20375.1 | |
| AK294313.1 | 5 | 506 | ||
| AK295366.1 | 6 | 603 | ||
| AL538140.3 | 6 | 615 | ||
| BC121802.1 | 5 | 544 | ||
| BC121803.1 | 6 | 751 |
More Information
Set Membership: |
|

Genomic Map