Assay Details
Target Gene Details
Entrez Gene ID: | 57644 |
Gene Name: | myosin heavy chain 7B |
Gene Aliases: |
MHC14, MYH14 |
Location: |
Chr.20:34955835-35002437 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 47 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYH7B | NM_020884.4 | NP_065935.3 | ||
| XM_006723840.3 | XP_006723903.1 | |||
| XM_011528941.2 | XP_011527243.1 | |||
| XM_011528947.2 | XP_011527249.1 | |||
| XM_011528948.2 | XP_011527250.1 | |||
| XM_017027986.1 | XP_016883475.1 | |||
| XM_017027987.1 | XP_016883476.1 | |||
| AB040945.2 | ||||
| BC007808.1 | AAH07808.1 | |||
| BC151242.1 | ||||
| BG189104.1 |
Target Gene Details
Entrez Gene ID: | 26133 |
Gene Name: | transient receptor potential cation channel subfamily C member 4 associated protein |
Gene Aliases: |
C20orf188, PPP1R158, TRRP4AP, TRUSS |
Location: |
Chr.20:35002404-35092837 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC4AP | NM_015638.2 | NP_056453.1 | ||
| NM_199368.1 | NP_955400.1 | |||
| XM_011528774.1 | XP_011527076.1 | |||
| XM_017027800.1 | XP_016883289.1 | |||
| XM_017027801.1 | XP_016883290.1 | |||
| AF055022.1 | ||||
| AK074106.1 | BAB84932.1 | |||
| AL096738.1 | ||||
| AL117480.1 | CAB55953.1 | |||
| BC001323.1 | AAH01323.1 | |||
| BC008836.2 | AAH08836.2 | |||
| BC013144.1 | AAH13144.1 | |||
| BM723903.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063673 | Chr.20:34977862 - 35009073 on Build GRCh38 | Loss |
|
| nsv1055447 | Chr.20:34737641 - 35020344 on Build GRCh38 | Loss |
|
| nsv833962 | Chr.20:34927688 - 35113250 on Build GRCh38 | Loss |
|
| nsv3362 | Chr.20:34978502 - 35010725 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map