Assay Details
Target Gene Details
Entrez Gene ID: | 23101 |
Gene Name: | MCF.2 cell line derived transforming sequence-like 2 |
Gene Aliases: |
ARHGEF22 |
Location: |
Chr.3:183176116-183428990 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCF2L2 | NM_015078.3 | 30 | 4242 | NP_055893.3 |
| XM_011512585.2 | 22 | 3679 | XP_011510887.1 | |
| XM_011512586.2 | 23 | 3093 | XP_011510888.1 | |
| AB020668.1 | 27 | 3554 | BAA74884.1 | |
| BC029074.2 | 8 | 1616 | AAH29074.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822362 | Chr.3:183178075 - 183180115 on Build GRCh38 | Gain |
|
| dgv960e214 | Chr.3:183134096 - 183206174 on Build GRCh38 | Gain |
|
| esv3893833 | Chr.3:183130677 - 183342208 on Build GRCh38 | Gain |
|
| nsv508970 | Chr.3:183126164 - 183213219 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map