Assay Details
Target Gene Details
Entrez Gene ID: | 55330 |
Gene Name: | biogenesis of lysosomal organelles complex 1 subunit 4 |
Gene Aliases: |
BCAS4L, BLOS4, CNO |
Location: |
Chr.4:6716115-6717660 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLOC1S4 | NM_018366.2 | 1 | 406 | NP_060836.1 |
| AK002092.1 | 1 | 406 | BAA92080.1 | |
| BC067815.1 | 1 | 347 | AAH67815.1 |
More Information
Set Membership: |
|

Genomic Map