Assay Details
Target Gene Details
Entrez Gene ID: | 91860 |
Gene Name: | calmodulin like 4 |
Gene Aliases: |
NY-BR-20 |
Location: |
Chr.15:68190705-68206110 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALML4 | NM_001031733.2 | 4 | 2645 | NP_001026903.2 |
| NM_001286694.1 | 4 | 2755 | NP_001273623.1 | |
| NM_001286695.1 | 3 | 2614 | NP_001273624.1 | |
| NM_033429.2 | 5 | 2786 | NP_219501.2 | |
| NR_104583.1 | 2 | 2425 | ||
| AK092628.1 | 4 | 1867 | ||
| BX640702.1 | 3 | 1803 | CAE45822.1 | |
| BX640817.1 | 2 | 2426 | CAE45894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1581 | Chr.15:68172970 - 68204223 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map