Assay Details
Target Gene Details
Entrez Gene ID: | 2965 |
Gene Name: | general transcription factor IIH subunit 1 |
Gene Aliases: |
BTF2, P62, TFB1, TFIIH |
Location: |
Chr.11:18322269-18367043 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF2H1 | NM_001142307.1 | NP_001135779.1 | ||
| NM_005316.3 | NP_005307.1 | |||
| XM_006718208.3 | XP_006718271.1 | |||
| AK027003.1 | BAB15621.1 | |||
| AK127204.1 | ||||
| AK299621.1 | ||||
| AK299730.1 | ||||
| AK312974.1 | ||||
| BC000365.2 | AAH00365.1 | |||
| BC004452.1 | AAH04452.1 | |||
| M95809.1 | AAA58399.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553633 | Chr.11:18290154 - 19036904 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map