Assay Details
Target Gene Details
Entrez Gene ID: | 27315 |
Gene Name: | post-GPI attachment to proteins 2 |
Gene Aliases: |
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21 |
Location: |
Chr.11:3797724-3826371 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv190n67 | Chr.11:3817510 - 3818529 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map