Assay Details
Target Gene Details
Entrez Gene ID: | 4915 |
Gene Name: | neurotrophic receptor tyrosine kinase 2 |
Gene Aliases: |
GP145-TrkB, TRKB, trk-B |
Location: |
Chr.9:84668368-85027070 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048977 | Chr.9:84149197 - 84815576 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map