Assay Details
Target Gene Details
Entrez Gene ID: | 100526737 |
Gene Name: | RBM14-RBM4 readthrough |
Gene Aliases: |
COAZ |
Location: |
Chr.11:66616582-66646473 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM14-RBM4 | NM_001198845.1 | 2 | 787 | NP_001185774.1 |
| NM_001198846.1 | NP_001185775.1 | |||
| AW958943.1 |
Target Gene Details
Entrez Gene ID: | 5936 |
Gene Name: | RNA binding motif protein 4 |
Gene Aliases: |
LARK, RBM4A, ZCCHC21, ZCRB3A |
Location: |
Chr.11:66638617-66668385 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM4 | NM_001198843.1 | NP_001185772.1 | ||
| NM_001198844.1 | NP_001185773.1 | |||
| NM_002896.3 | 3 | 871 | NP_002887.2 | |
| AK097592.1 | 2 | 1500 | ||
| AK225071.1 | 3 | 785 | ||
| AK303984.1 | ||||
| BC000307.2 | 3 | 769 | AAH00307.1 | |
| BC032735.1 | 3 | 792 | AAH32735.1 | |
| BC064960.1 | AAH64960.1 | |||
| U89505.1 | 3 | 778 | AAC51293.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518919 | Chr.11:66641938 - 66700998 on Build GRCh38 | Loss |
|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map