Assay Details
Target Gene Details
Entrez Gene ID: | 285242 |
Gene Name: | 5-hydroxytryptamine receptor 3E |
Gene Aliases: |
5-HT3-E, 5-HT3E, 5-HT3c1 |
Location: |
Chr.3:184097064-184106995 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR3E | NM_001256613.1 | NP_001243542.1 | ||
| NM_001256614.1 | 2 | 596 | NP_001243543.1 | |
| NM_182589.2 | NP_872395.2 | |||
| NM_198313.2 | NP_938055.1 | |||
| NM_198314.2 | NP_938056.1 | |||
| AY159813.2 | ||||
| AY349352.1 | AAQ93476.1 | |||
| AY349353.1 | AAQ93477.1 | |||
| BC101182.1 | AAI01183.1 | |||
| BC101183.2 | AAI01184.1 | |||
| BC101185.2 | AAI01186.1 | |||
| DQ644022.1 | ||||
| EU165354.1 | 2 | 402 |
More Information
Set Membership: |
|

Genomic Map