Assay Details
Target Gene Details
Entrez Gene ID: | 4351 |
Gene Name: | mannose phosphate isomerase |
Gene Aliases: |
CDG1B, PMI, PMI1 |
Location: |
Chr.15:74890011-74899457 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPI | NM_001289155.1 | 3 | 296 | NP_001276084.1 |
| NM_001289156.1 | 2 | 168 | NP_001276085.1 | |
| NM_001289157.1 | 3 | 296 | NP_001276086.1 | |
| NM_002435.2 | 3 | 296 | NP_002426.1 | |
| XM_011521592.1 | 3 | 372 | XP_011519894.1 | |
| XM_011521593.2 | 3 | 354 | XP_011519895.1 | |
| XM_017022207.1 | 3 | 376 | XP_016877696.1 | |
| XM_017022208.1 | 3 | 354 | XP_016877697.1 | |
| XM_017022209.1 | 2 | 143 | XP_016877698.1 | |
| AF504648.1 | 3 | 261 | AAM28199.1 | |
| AK225149.1 | 3 | 259 | ||
| AK292374.1 | 3 | 249 | ||
| AK294030.1 | 3 | 265 | ||
| AK301370.1 | 2 | 137 | ||
| AK302354.1 | 2 | 137 | ||
| BC017351.2 | 3 | 245 | AAH17351.1 | |
| BC046357.1 | 3 | 251 | AAH46357.1 | |
| BX648475.1 | 3 | 273 | ||
| CD706167.1 | 3 | 287 | ||
| X76057.1 | 3 | 238 | CAA53657.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv990390 | Chr.15:74886959 - 74895604 on Build GRCh38 | Deletion |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map